Product Name
MISSION® esiRNA targeting mouse Dstn
Product Number
EMU054131
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
Appearancecolorless liquid
Concentration200 ng/µL
PCR Product - Gel ElectrophoresisPass
PCR Product - DNA SequencingPass
Concentration - OD260 or Fluorescence IntensityPass
Purified RNaseIII - Gel ElectrophoresisPass