Product Name
Gemfibrozil
Product Number
G9518
Product Brand
SIGMA
CAS Number
25812-30-0
Molecular Weight
250.33
Test
Specification
Appearance (Color)White to Faint Yellow
Appearance (Form)Powder
Solubility (Color)Colorless
Solubility (Turbidity)Clear
 50 mg/mL, EtOH
Carbon70.2 - 73.8 %
Purity (GC)≥99 %