Product Name
Human Serum from clotted human male whole blood, sterile-filtered, (mycoplasma tested, virus tested)
Product Number
H1388
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
SOURCEMALE DONOR'S WITHIN THE UNITED STATES
PROCESSING COUNTRY OF ORIGINUNITED STATES
APPEARANCEHAZY STRAW TO AMBER COLORED LIQUID
STERILITY TESTSTERILE
MYCOPLASMA TEST BY BARILENONE DETECTED
HEMOGLOBIN ASSAYNMT 20 MG%
ENDOTOXIN ASSAYNMT 10 EU/ML
OSMOLALITY260 - 340 MOSM/KG WATER
PH TEST7.0 - 8.0
CHOLESTEROLREPORT RESULT
TRIGLYCERIDEREPORT RESULT
GLUCOSEREPORT RESULT
SODIUMREPORT RESULT
IRONREPORT RESULT
PROTEIN CONTENT4.0 - 9.0 G%
NOTEEACH DONOR UNIT USED IN THE PREPARATION OF THE PRODUCT WAS TESTED AND FOUND NEGATIVE FOR HBSAG, ANTI-HCV AND ANTI-HIV/HIV-2 BY FDA APPROVED TEST.
ALL DONOR UNITS ARE COLLECTED IN DONOR CENTERS LOCATED IN THE UNITED STATES, WHICH ARE LICENSED BY THE FDA.
ALL BLOOD PRODUCTS SHOULD BE TREATED AND HANDLED AS POTENTIALLY INFECTIOUS.
DOCUMENT # H1388:04/01-7