Product Name
Hexamethyldisilazane
Product Number
H4875
Product Brand
SIGMA
CAS Number
999-97-3
Molecular Weight
161.39
Test
Specification
Appearance (Color)Colorless
Appearance (Form)Liquid
Appearance (Turbidity)Clear
Proton NMR spectrumConforms to Structure
Purity (GC)≥97 %