Product Name
Anti-ZNF497 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Product Number
HPA047872
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
Protein Arraypass
Immunohistochemistry Dilution1:50 - 1:200
Immunohistochemistry Antibody Retrieval MethodHIER pH6
Concentration0.1 mg/mL
Western Blot ScoreSingle band corresponding to predicted size in kDa (+/- 20%)