Product Name
Anti-ZNF334 antibody produced in rabbit Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Product Number
HPA050022
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
Protein Arraypass
Immunohistochemistry Dilution1:200 - 1:500
Immunohistochemistry Antibody Retrieval MethodHIER pH6
Concentration0.1 mg/mL