Product Name
Ibotenic acid ~95%, solid
Product Number
I2765
Product Brand
SIGMA
CAS Number
2552-55-8
Molecular Weight
158.11
Test
Specification
APPEARANCEWHITE TO LIGHT YELLOW POWDER
SOLUBILITYCLEAR COLORLESS TO YELLOW SOLUTION AT 50 MG/ML IN 1 N AMMONIUM HYDROXIDE
IR SPECTRUMCONSISTENT WITH STRUCTURE
WATER BY KARL FISCHERNMT 10%
ELEMENTAL ANALYSIS *35.0 TO 40.0% CARBON
16.3 TO 18.6% NITROGEN
PURITY BY THIN LAYER CHROMATOGRAPHYAPPROX. 95%
* ANHYDROUS BASIS
APPROVED JLH 2/9/95