Product Name
3-Methylbutanol ~98%
Product Number
I3643
Product Brand
SIGMA
CAS Number
123-51-3
Molecular Weight
88.15
Test
Specification
APPEARANCECLEAR COLORLESS LIQUID
IR SPECTRUMCONSISTENT WITH STRUCTURE
PURITY BY GAS CHROMATOGRAPHYMINIMUM 98%
SHELF LIFE2 YEARS
DOCUMENT # I3643/06/24/99/1