Product Name
Sodium dodecyl sulfate for molecular biology, ≥98.5% (GC)
Product Number
L4390
Product Brand
SIGMA
CAS Number
151-21-3
Molecular Weight
288.38
Test
Specification
APPEARANCEWHITE TO OFF-WHITE POWDER
SOLUBILITYCLEAR COLORLESS TO VERY FAINT YELLOW SOLUTION AT 20G PLUS 100ML OF WATER
WATER BY KARL FISCHER< OR = 2.0%
ELEMENTAL ANALYSIS7.0 TO 9.0 % SODIUM
ELEMENTAL ANALYSIS< OR = 10 PPM LEAD
ELEMENTAL ANALYSIS< OR = 500 PPM CHLORIDE
PHOSPHATE< OR = 10 PPM
UV ABSORBANCE (3% W/V IN WATER)< OR = 0.3 AT 260 NM
UV ABSORBANCE (3% W/V IN WATER)< OR = 0.3 AT 280 NM
DNASE, (EXONUCLEASE)NONE DETECTED
RNASENONE DETECTED
PURITY BY TITRATION> OR = 98.5%
PURITY BY GAS CHROMATOGRAPHY> OR = 98.5%
RECOMMENDED RETEST3 YEARS
DOCUMENT #L4390/08/10/09/3