Product Name
N-(Methoxysuccinyl)-Ala-Ala-Pro-Val-chloromethyl ketone elastase inhibitor
Product Number
M0398
Product Brand
SIGMA
CAS Number
65144-34-5
Molecular Weight
502.99
Test
Specification
APPEARANCEWHITE POWDER
SOLUBILITYCLEAR COLORLESS SOLUTION AT 50 MG/ML IN METHANOL
CARBON51.2 TO 53.8%
NITROGEN10.8 TO 11.4%
PURITY BY THIN LAYER CHROMATOGRAPHY>OR = 98%
DOCUMENT #M0398/11/04/08/1