Product Name
6α-Methylprednisolone ≥98%
Product Number
M0639
Product Brand
SIGMA
CAS Number
83-43-2
Molecular Weight
374.47
Test
Specification
APPEARANCEWHITE TO OFF-WHITE POWDER
SOLUBILITYCLEAR COLORLESS TO FAINT YELLOW SOLUTION AT 50MG/ML IN CHLOROFORM:METHANOL (1:1)
ULTRAVIOLET/VISIBLE SPECTRUMEMM = 13.8 TO 14.7 AT LAMBDA MAX 241 TO 245NM IN ETHANOL
PURITY BY HPLC> OR = 98%
RECOMMENDED RETEST3 YEARS
DOCUMENT #M0639/08/19/10/5