Product Name
MEM Non-essential Amino Acid Solution (100×) without L-glutamine, liquid, sterile-filtered, BioReagent, suitable for cell culture
Product Number
M7145
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
Appearance (Turbidity)Clear
Appearance (Form)Solution
pH1.4 - 2.0
Osmolality- mOs/kg
SterilityPass
Endotoxin at 30mL/L<= 0.5 EU/ml
Cell Culture Testing - MTTPass
Cell LineCell Line - Cell Types
Animal Derived Component FreeConforms