Product Name
5β-Pregnane-3α,20α-diol glucuronide
Product Number
P3635
Product Brand
SIGMA
CAS Number
1852-49-9
Molecular Weight
496.63
Test
Specification
APPEARANCEWHITE POWDER
SOLUBILITYCLEAR COLORLESS SOLUTION AT 20MG PLUS 1 ML OF ETHANOL
IR SPECTRUMCONSISTENT WITH STRUCTURE
WATER BY KARL FISCHER< OR = 10%
CARBON63.7 TO 66.9% *
PURITY BY THIN LAYER CHROMATOGRAPHY> OR = 98%
*ANHYDROUS BASIS
DOCUMENT #P3635/02/09/09/1