Product Name
Protein Kinase G Iβ human ≥95% (SDS-PAGE), recombinant, expressed in baculovirus infected Sf9 cells, buffered aqueous glycerol solution
Product Number
P3738
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
µg/ml≥25
 Protein Content
units/mg protein≥1.5
 One unit will phosphorylate 1.0 micromole of
 VASPtide (RRKVSKQE) substrate at pH 7.4 at 30 deg
 C
Purity by SDS ElectrophoresisConforms
 >= 95%