Product Name
Anti-Presenilin-1 (14-33) antibody produced in goat whole antiserum
Product Number
P5235
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
Miscellaneous Supplier DataConforms
 1:1000 on frozen sections (recommended dilution -
 optimal working dilutions should be determined by
 titration test)