Product Name
Protein Kinase A from bovine heart ≥0.4 units/μg protein, lyophilized powder
Product Number
P5511
Product Brand
SIGMA
CAS Number
9026-43-1
Molecular Weight
Test
Specification
PROTEIN BY BIURET> OR = 70%
ENZYMATIC ACTIVITY> OR = 0.4 UNITS/UG PROTEIN
UNIT DEFINITIONONE UNIT WILL TRANSFER 1 PICOMOLE OF PHOSPHATE FROM GAMMA-32P-ATP TO HYDROLYZED AND PARTIALLY DEPHOSPHORYL- ATED CASEIN, PER MINUTE AT PH 6.5 AT 30 DEG C IN THE PRESENCE OF CYCLIC-AMP.
DOCUMENT #P5511/02/17/11/3