Product Name
Phospholipase D from cabbage Type IV, lyophilized powder, ≥100 units/mg solid
Product Number
P8398
Product Brand
SIGMA
CAS Number
9001-87-0
Molecular Weight
Test
Specification
APPEARANCEFAINT GREEN TO BROWN POWDER
PROTEIN BY BIURET> OR = 20%
ENZYMATIC ACTIVITY> OR = 100 UNITS/MG SOLID
UNIT DEFINITIONONE UNIT WILL LIBERATE 1.0MICROMOLE OF CHOLINE FROM L-ALPHA-PHOSPATIDYL- CHOLINE (EGG YOLK) PER HOUR AT PH5.6 AT 30DEGC.
DOCUMENT # P8398/10/19/10/3