Product Name
TruPAGE TEA-Tricine SDS Running Buffer 20 ×
Product Number
PCG3001
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
pH at 25 deg C8.30-8.60
Conductivity7.0-8.0mS
Refractive Index29.0-32.0 deg.Brix
Functional testPass
Overall resultPass
Expiration date2 years from date of manufacture