Product Name
TruPAGE Tris-MOPS SDS Express Running Buffer 20 ×
Product Number
PCG3003
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
pH at 25 deg C7.80-8.10
Conductivity12.1-12.7mS
Refractive Index31.0-32.0deg.Brix
Functional testReport result
Overall resultReport result
Expiration date2 years from date of manufacture