Product Name
Anti-FRAT1 (C-term) antibody produced in rabbit saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution
Product Number
SAB1300183
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
Appearancecolorless liquid
Concentration>= 0.25 mg/ml
ELISA1:1000
Western Blotting1:100-1:500
Immunohistochemistry1:50-1:100
ImmunofluorescenceNot Verified
ImmunoprecipitationNot Verified
Flow CytometryNot Verified
Chromatin ImmunoprecipitationNot Verified