Product Name
Anti-CYP27A1 (N-term) antibody produced in rabbit saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution
Product Number
SAB1300712
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
Appearancecolorless liquid
Concentration>= 0.25 mg/ml
ELISA1:1000
Western Blotting1:100-1:500
ImmunohistochemistryNot Verified
ImmunofluorescenceNot Verified
ImmunoprecipitationNot Verified
Flow CytometryNot Verified
Chromatin ImmunoprecipitationNot Verified