Product Name
Anti-SPP (N-term) antibody produced in rabbit IgG fraction of antiserum, buffered aqueous solution
Product Number
SAB1300769
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
Appearancecolorless liquid
Concentration>= 0.25 mg/ml
ELISA1:1000
Western BlottingNot Verified
ImmunohistochemistryNot Verified
ImmunofluorescenceNot Verified
ImmunoprecipitationNot Verified
Flow CytometryNot Verified
Chromatin ImmunoprecipitationNot Verified