Product Name
Monoclonal Anti-NR4A3 antibody produced in mouse clone 1E9, purified immunoglobulin, buffered aqueous solution
Product Number
SAB1404566
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
AppearanceReport result
ConcentrationReport result
Western Blot(Cell lysate)Antibody concentration:1~5µg/mL.Specific band of ~68.2kDa using Hela NE lysate.
Immunofluorescence(IF)Antibody concentration:10µg/mL using Hela cell.
Sandwich ELISADetection limit for recombinant GST tagged NR4A3 is approximately 0.3ng/mL.
ELISA(Recombinant protein)Antibody concentration:1µg/mLusing recombinant protein.
Western Blotting(Recombinant protein)Antibody concentration:1µg/mL.Specific band of ~38kDa using immunogen protein lysate.