Product Name
ANTI-HOXA5 antibody produced in mouse clone 4E11, purified immunoglobulin, buffered aqueous solution
Product Number
SAB1412464
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
Appearancecolorless liquid
ConcentrationReport Result
Storage/StabilityReport Result
Storage Temperature-20�C
Species ReactivityReport Result
ELISAAntibody concentration: 1 ug/ml using recombinant protein.
Sandwich ELISADetection limit for recombinant GST tagged HOXA5 is approximately 0.3ng/ml.
Western Blotting (Recombinant Protein)Antibody concentration: 1 ug/ml. Specific band of ~36.6 kDa using immunogen protein lysate.
Western Blotting (Cell Lysate)Not Verified
Western Blotting (Tissue lysate)Not Verified
Western Blotting (Transfected lysate)Not Verified
ImmunohistochemistryNot Verified
ImmunofluorescenceAntibody concentration: 10 ug/ml using HeLa cell.
HandlingReport Result