Product Name
Anti-phospho-MAP2K1 (pSer221) antibody produced in rabbit affinity isolated antibody
Product Number
SAB4300132
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
Appearancecolorless liquid
Concentration1.0 mg/ml
ELISANot Determined
Western Blotting1:500-1:1000
Immunohistochemistry1:50-1:100
Immunofluorescence1:100-1:200
ImmunoprecipitationNot Determined
Flow CytometryNot Determined
Chromatin ImmunoprecipitationNot Determined