Product Name
Anti-STAT5A (Ab-780) antibody produced in rabbit affinity isolated antibody
Product Number
SAB4300330
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
Appearancecolorless liquid
Concentration1.0 mg/ml
ELISANot Determined
Western Blotting1:500-1:1000
Immunohistochemistry1:50-1:100
ImmunofluorescenceNot Determined
ImmunoprecipitationNot Determined
Flow CytometryNot Determined
Chromatin ImmunoprecipitationNot Determined