Product Name
Anti-phospho-VEGFR1 (pTyr1333) antibody produced in rabbit affinity isolated antibody
Product Number
SAB4504006
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
AppearanceReport result
ConcentrationReport result
PurificationAffinity purified
ELISA1:40000
Western Blotting1:500-1:1000
Immunohistochemistry1:50-1:100