Product Name
Caspase 13 Substrate
Product Number
SCP0074
Product Brand
SIGMA
CAS Number
Molecular Weight
703.69
Test
Specification
Formlyophilized powder
AppearanceWhite to off-white
Peptide Content> or = to 65%
Purity by HPLC> or = to 95%