Product Name
Caspase 13 Substrate
Product Number
SCP0075
Product Brand
SIGMA
CAS Number
Molecular Weight
666.63
Test
Specification
Formlyophilized powder
AppearanceWhite to off-white
Peptide Content> or = to 80%
Purity by HPLC> or = to 95%