Product Name
Protein Kinase C Substrate
Product Number
SCP0227
Product Brand
SIGMA
CAS Number
Molecular Weight
1197.48
Test
Specification
Formlyophilized powder
AppearanceWhite to off-white
Peptide Content> or = to 65%
Purity by HPLC> or = to 95%