Product Name
Neurturin human recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Product Number
SRP3124
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
AppearanceReport result
ColorReport result
Purity by SDS-PAGE≥98%
Purity by HPLC≥98%
Endotoxin level≤1 EU/µg
Biological ActivityHuman Neurturin at a concentration of 100 ng/ml will support the survival of 65% of newborn rat sympathetic neurons.