Product Name
Persephin human recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Product Number
SRP3141
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
AppearanceReport result
ColorReport result
Purity by SDS-PAGE≥98%
Purity by HPLC≥98%
Endotoxin Level≤1 EU/µg
Biological ActivityHuman Persephin induces RET phosphorylation using a concentration range of 0.1-1.0 ng/ml. Human Persephin binds to mammalian GFRa4 with the Kd of 100pM. Other members of the GDNF family (Artemin, GDNF and Neurturin) do not bind to mammalian GFRa4.