Product Name
VEGF human Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Product Number
SRP3182
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
FormReport result
Purity≥95%
Endotoxin≤ 1 EUs/µg
Biological ActivityED50 ≤ 10 ng/mL (≥ 1.0 x 105 units/mg)