Product Name
NOGGIN from mouse recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Product Number
SRP3227
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
FormReport result
ColorReport result
Purity by SDS Page≥98%
Purity by HPLC≥98%
Endotoxin Level≤1 EU/µg
Biological ActivityDetermined by its ability to inhibit 5.0ng/ml of BMP-4 induced alkaline phosphatase production by ATDC chondrogenic cells. The expected ED50 for this effect is 1.0-2.0 ng/ml of Noggin