Product Name
SCF from rat recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Product Number
SRP3251
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
Appearancelyophilized powder
Colorwhite to off-white
Purity by SDS-Page>=98%
Purity by HPLC>=98%
Endotoxin Level<= 1 EU/ug
Biological ActivityDetermined by the dose-dependent stimulation of the proliferation of human TF-1 cells. The expected ED50 for this effect is 20-40 ng/ml.