Product Name
IFN-alpha 1 human recombinant, expressed in E. coli, ≥95% (SDS-PAGE), ≥95% (HPLC)
Product Number
SRP4596
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
AppearanceReport result
Purity by SDS-PAGE≥ 95 %
Purity by HPLC≥ 95 %
Endotoxin< 0.1 ng/ µg protein by LAL Method
Biological ActivityIFN-alpha-1 was titrated using a cytopathic effect inhibition assay