Product Name
Ubiquitin Activating Enzyme E1 human ≥95% (SDS-PAGE)
Product Number
SRP6147
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
Purity by SDS-PAGE≥ 95 %
ActivityTypical enzyme concentration of 5 nM 100 nM is used for in vitro conjugation, depending on conditions.