Product Name
Trizma® hemisulfate
Product Number
T8379
Product Brand
SIGMA
CAS Number
6992-38-7
Molecular Weight
170.17
Test
Specification
APPEARANCEWHITE POWDER
SOLUBILITYCLEAR COLORLESS SOLUTION AT 25 GM PLUS 50 ML OF WATER
WATER BY KARL FISCHERNMT 0.5%
IR SPECTRUMCONSISTENT WITH STRUCTURE
HEAVY METALSNMT 5 PPM AS LEAD
UV ABSORBANCENMT 0.05 AT 290 NM (33% W/W IN WATER)
PURITY BY SODIUM HYDROXIDE TITRATIONMINIMUM 99%
DOCUMENT #T8379/09/20/06/1