Product Name
Trizma® succinate
Product Number
T9632
Product Brand
SIGMA
CAS Number
85169-32-0
Molecular Weight
360.36
Test
Specification
APPEARANCEWHITE CRYSTALLINE POWDER
SOLUBILITYCLEAR COLORLESS SOLUTION AT 1G/ML IN WATER
WATER BY KARL FISCHERNMT 0.1%
HEAVY METALSNMT 5 PPM (ASLEAD)
UV ABSORBANCENMT 0.1 AT A290 NM (50%W/W)
PURITY BY TITRATIONMINIMUM 98%
DOCUMENT # T9632/11/24/98/1