Product Name
4-Imidazoleacrylic acid ≥99%
Product Number
U7500
Product Brand
SIGMA
CAS Number
104-98-3
Molecular Weight
138.12
Test
Specification
APPEARANCEWHITE TO OFF-WHITE POWDER
SOLUBILITYCLEAR TO SLIGHTLY HAZY COLORLESS TO FAINT YELLOW SOLUTION AT 50MG/ML IN 0.5M HYDROCHLORIC ACID
PURITY BY TITRATIONNLT 99%
PURITY BY THIN LAYER CHROMATOGRAPHYNLT 99%
DOCUMENT # U7500/11/02/00/5