Product Name
Williams’ Medium E With sodium bicarbonate, without L-glutamine and phenol red, liquid, sterile-filtered, suitable for cell culture
Product Number
W1878
Product Brand
SIGMA
CAS Number
Molecular Weight
Test
Specification
Appearance (Turbidity)Clear
Appearance (Form)Solution
pH7.0 - 7.6
Osmolality285 - 315 mOs/kg
Cell Culture Testing - MTTPass
Cell LineCell Line - Cell Types
SterilityPass
Endotoxin Level<= 1 EU/ml
Glucose Concentration1.8 - 2.2 g/l