Product Name
Zymosan A from Saccharomyces cerevisiae for inducing inflamatory response
Product Number
Z4250
Product Brand
SIGMA
CAS Number
58856-93-2
Molecular Weight
Test
Specification
APPEARANCEOFF-WHITE TO LIGHT BROWNISH POWDER
LOSS ON DRYING< OR = 10%
ELEMENTAL ANALYSIS *1.5 TO 4.0% NITROGEN
PHOSPHORUS *0.4 TO 1.4%
* DRY BASIS
DOCUMENT #Z4250/01/28/09/3