Product Name
Zinc chloride ≥98%
Product Number
Z4875
Product Brand
SIGMA
CAS Number
7646-85-7
Molecular Weight
136.30
Test
Specification
APPEARANCEWHITE TO OFF-WHITE POWDER
SOLUBILITYCLEAR TO VERY SLIGHTLY HAZY COLORLESS TO FAINT YELLOW SOLUTION WITH SOME INSOLUBLES AT 20G IN 200ML OF WATER PLUS 6ML 1M HYDROCHLORIC ACID
PURITY BY SILVER NITRATE TITRATIONMINIMUM 97%
RECOMMENDED RETEST4 YEARS
DOCUMENT # Z4875/03/24/00/2