Product Name
Diheptyl phthalate analytical standard
Product Number
00282
Product Brand
SUPELCO
CAS Number
3648-21-3
Molecular Weight
362.50
Test
Specification
APPEARANCE (COLOR)Colorless to Yellow
APPEARANCE (FORM)Liquid
PURITY (GC AREA %)≥ 97.0 %
CARBON CONTENT72.89 % (theory)
HYDROGEN CONTENT9.45 % (theory)
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS