Product Name
NTA - Atto 647 N BioReagent, suitable for fluorescence, ≥80% (HPCE)
Product Number
02175
Product Brand
SUPELCO
CAS Number
Molecular Weight
Test
Specification
APPEARANCE (COLOR)Dark Blue to Very Dark Blue
APPEARANCE (FORM)Powder or Film
PURITY (HPCE AREA %)≥ 80.0 %
SOLUBILITY (COLOR)REPORT RESULT
SOLUBILITY (TURBIDITY)REPORT RESULT
SOLUBILITY (METHOD)20 UL/VIAL in Water
EMISSION MAXIMUM669 ± 5 nm
REMARKS ON LUMINESCENCEEX. 647 NM IN PBS