Product Name
N-Acetylglycine analytical standard
Product Number
69563
Product Brand
SUPELCO
CAS Number
543-24-8
Molecular Weight
117.10
Test
Specification
APPEARANCE (COLOR)White
APPEARANCE (FORM)Powder or Crystals
TITRATION (T) NAOH 0.1M98.0 - 102.0 %
PURITY (HPLC AREA %)≥ 95 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS