Product Name
1,3,5-Tri-tert-butylbenzene analytical standard
Product Number
94056
Product Brand
SUPELCO
CAS Number
1460-02-2
Molecular Weight
246.43
Test
Specification
APPEARANCE (COLOR)White to Off White
APPEARANCE (FORM)Powder or Crystals or Fibers
PURITY (GC AREA %)≥ 98.0 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE
EXPIRY PERIOD36 MONTHS