Product Name
Acetylacetone produced by Wacker Chemie AG, Burghausen, Germany, ≥99.5% (GC)
Product Number
10916
Product Brand
aldrich
CAS Number
123-54-6
Molecular Weight
100.12
Test
Specification
APPEARANCE (COLOR)Colorless
APPEARANCE (FORM)Liquid
APPEARANCE (TURBIDITY)Clear
COLOR (IN APHA)≤ 20 APHA
PURITY (GC AREA %)≥ 99.5 %
REMARKS ON GCOTHER SINGLE IMPURITIES ≤ 0.1 %
WATER≤ 0.1 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE
ACIDITYACETIC ACID ≤ 0.05 %
VENDOR INFORMATIONProduct of Wacker