Product Name
4-Bromobenzyl bromide 98%
Product Number
112186
Product Brand
aldrich
CAS Number
589-15-1
Molecular Weight
249.93
Test
Specification
APPEARANCE (COLOR)White to Light Yellow and Faint Orange to Light Orange and Faint Beige to Light Beige and Faint Brown to Light Brown
APPEARANCE (FORM)Powder or Crystals or Flakes or Chunks
PURITY (GC AREA %)≥ 97.5 %
PROTON NMR SPECTRUMCONFORMS TO STRUCTURE