Product Name
Mowiol® 20-98 Mw ~125,000
Product Number
11773
Product Brand
aldrich
CAS Number
9002-89-5
Molecular Weight
Test
Specification
APPEARANCE (COLOR)Colorless or White
APPEARANCE (FORM)Powder or Crystals or Flakes
RESIDUE ON IGNITION≤ 0.5 %
INFRARED SPECTRUMCONFORMS TO STRUCTURE
VISCOSITY (ROTATION)18.5 - 21.5 mPas
VISCOSITY (CONDITIONS)4 % IN H2O, 20 C
MISCELLANEOUS TESTSDEGREE OF HYDROLYSIS 98.0-98.8 % MOL